COMH93D5C6C6 Combi-Blocks, Inc.

Silicon tetraisothiocyanate

  • MDL number MFCD00054206
  • Cas Number : 6544-02-1
  • Molecular Weight : 260.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6238

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6238
Availability details for
Quantity
  • Availability
  • Add to Cart