COMH93D5C6CC Combi-Blocks, Inc.

1-(3-Chlorophenoxymethyl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD07995738
  • Molecular Weight : 234.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6246
Availability details for
Quantity
  • Availability
  • Add to Cart