COMH93D5C6DD Combi-Blocks, Inc.

Methyl 7-oxo-7h-thiazolo[3,2-a]pyrimidine-5-carboxylate

  • MDL number MFCD00277522
  • Cas Number : 33304-83-5
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6274
Availability details for
Quantity
  • Availability
  • Add to Cart