COMH93D5C77E Combi-Blocks, Inc.

Tetraethylammonium trifluoroacetate

  • MDL number MFCD00192470
  • Cas Number : 30093-29-9
  • Molecular Weight : 243.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6518

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6518
Availability details for
Quantity
  • Availability
  • Add to Cart