COMH93D5C835 Combi-Blocks, Inc.

Ethyl 6-methoxynicotinate

  • MDL number MFCD06204771
  • Cas Number : 74925-37-4
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6788
Availability details for
Quantity
  • Availability
  • Add to Cart