COMH93D5C8D4 Combi-Blocks, Inc.

4,6-Dichloropyridine-3-acetic acid

  • MDL number MFCD04114264
  • Cas Number : 199283-51-7
  • Molecular Weight : 206.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7025
Availability details for
Quantity
  • Availability
  • Add to Cart