COMH93D5C8DB Combi-Blocks, Inc.

Dihydrostreptomycin

  • MDL number MFCD00084778
  • Cas Number : 128-46-1
  • Molecular Weight : 583.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7036
Availability details for
Quantity
  • Availability
  • Add to Cart