COMH93D5C8E6 Combi-Blocks, Inc.

Z-Asp-2,6-dichlorobenzoyloxymethylketone

  • MDL number MFCD00674885
  • Cas Number : 153088-73-4
  • Molecular Weight : 454.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7053
Availability details for
Quantity
  • Availability
  • Add to Cart