COMH93D5C90B Combi-Blocks, Inc.

2-Isopropoxy-5-nitrobenzoic acid

  • MDL number MFCD02670138
  • Cas Number : 166263-28-1
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7105
Availability details for
Quantity
  • Availability
  • Add to Cart