COMH93D5C919 Combi-Blocks, Inc.

Ethyl [(2-methoxyphenyl)amino](oxo)acetate

  • MDL number MFCD00966766
  • Cas Number : 7267-26-7
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7120
Availability details for
Quantity
  • Availability
  • Add to Cart