COMH93D5C91E Combi-Blocks, Inc.

Tridecyl methacrylate

  • MDL number MFCD00059220
  • Cas Number : 2495-25-2
  • Molecular Weight : 268.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7129

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7129
Availability details for
Quantity
  • Availability
  • Add to Cart