COMH93D5C924 Combi-Blocks, Inc.

4-Piperazin-1-yl-thieno[3,2-c]pyridine

  • MDL number MFCD06796184
  • Cas Number : 106261-27-2
  • Molecular Weight : 219.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7143
Availability details for
Quantity
  • Availability
  • Add to Cart