COMH93D5C934 Combi-Blocks, Inc.

Cafenstrole

  • MDL number MFCD01631150
  • Cas Number : 125306-83-4
  • Molecular Weight : 350.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7169
Availability details for
Quantity
  • Availability
  • Add to Cart