COMH93D5C93C Combi-Blocks, Inc.

H-Leu-val-leu-ala-pna

  • MDL number MFCD02259654
  • Cas Number : 197970-37-9
  • Molecular Weight : 534.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7178

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7178
Availability details for
Quantity
  • Availability
  • Add to Cart