COMH93D5C960 Combi-Blocks, Inc.

Methyl 4-(1-amino-2-((tert-butoxycarbonyl)amino)ethyl)benzoate

  • MDL number MFCD09026588
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7224
Availability details for
Quantity
  • Availability
  • Add to Cart