COMH93D5C96D Combi-Blocks, Inc.

(3-Nitro-1h-pyrazol-1-yl)methanol

  • MDL number MFCD05667183
  • Cas Number : 1001419-82-4
  • Molecular Weight : 143.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7250
Availability details for
Quantity
  • Availability
  • Add to Cart