COMH93D5C9A6 Combi-Blocks, Inc.

(4-Bromophenyl)-(4-bromophenyl)imino-oxidoazanium

  • MDL number MFCD03940272
  • Cas Number : 1215-42-5
  • Molecular Weight : 356.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7330
Availability details for
Quantity
  • Availability
  • Add to Cart