COMH93D5CA37 Combi-Blocks, Inc.

Phenyl(pyridin-2-yl)methanol

  • MDL number MFCD00186514
  • Cas Number : 14159-57-0
  • Molecular Weight : 185.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7548
Availability details for
Quantity
  • Availability
  • Add to Cart