COMH93D5CA43 Combi-Blocks, Inc.

2-Methyl-7-phenyl-1h-indene

  • MDL number MFCD00674042
  • Cas Number : 153733-75-6
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7568
Availability details for
Quantity
  • Availability
  • Add to Cart