COMH93D5CA45 Combi-Blocks, Inc.

Meosuc-lys(2-picolinoyl)-ala-pro-val-pna

  • MDL number MFCD00238303
  • Cas Number : 757139-27-8
  • Molecular Weight : 752.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7572
Availability details for
Quantity
  • Availability
  • Add to Cart