COMH93D5CA4B Combi-Blocks, Inc.

2,3-Dimethoxy-5-iodobenzaldehyde

  • MDL number MFCD00016602
  • Cas Number : 7396-66-9
  • Molecular Weight : 292.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7579
Availability details for
Quantity
  • Availability
  • Add to Cart