COMH93D5CA5C Combi-Blocks, Inc.

9,9-Bis(4-methoxyphenyl)-9h-fluorene

  • MDL number MFCD00192781
  • Cas Number : 117766-40-2
  • Molecular Weight : 378.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7603
Availability details for
Quantity
  • Availability
  • Add to Cart