COMH93D5CA7A Combi-Blocks, Inc.

4,5,7-Trimethylcoumarin

  • MDL number MFCD01861448
  • Cas Number : 14002-91-6
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7649
Availability details for
Quantity
  • Availability
  • Add to Cart