COMH93D5CAAA Combi-Blocks, Inc.

N,N'-Ethylenebis(iodoacetamide)

  • MDL number MFCD00728185
  • Cas Number : 7250-43-3
  • Molecular Weight : 395.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7714

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7714
Availability details for
Quantity
  • Availability
  • Add to Cart