COMH93D5CAD8 Combi-Blocks, Inc.

[4-(Pyridin-3-yl)phenyl]methanamine

  • MDL number MFCD08544239
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7789
Availability details for
Quantity
  • Availability
  • Add to Cart