COMH93D5CADF Combi-Blocks, Inc.

2-(3-Bromo-phenyl)-isoindole-1,3-dione

  • MDL number MFCD01026470
  • Cas Number : 19357-22-3
  • Molecular Weight : 302.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7802

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7802
Availability details for
Quantity
  • Availability
  • Add to Cart