COMH93D5CAF3 Combi-Blocks, Inc.

2-(4-Chloro-phenyl)-benzooxazole

  • MDL number MFCD00453917
  • Cas Number : 1141-35-1
  • Molecular Weight : 229.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7828
Availability details for
Quantity
  • Availability
  • Add to Cart