COMH93D5CAFF Combi-Blocks, Inc.

Ginsenoside rh2

  • MDL number MFCD00800712
  • Cas Number : 67400-17-3
  • Molecular Weight : 622.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7851

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7851
Availability details for
Quantity
  • Availability
  • Add to Cart