COMH93D5CBF4 Combi-Blocks, Inc.

Bepridil

  • MDL number MFCD00868313
  • Cas Number : 64706-54-3
  • Molecular Weight : 366.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8233

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8233
Availability details for
Quantity
  • Availability
  • Add to Cart