COMH93D5CD57 Combi-Blocks, Inc.

Direct blue 6

  • MDL number MFCD00068426
  • Cas Number : 2602-46-2
  • Molecular Weight : 932.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8770
Availability details for
Quantity
  • Availability
  • Add to Cart