COMH93D5CD6A Combi-Blocks, Inc.

N-(5-Chloro-2-methyl-phenyl)-4-methyl-benzenesulfonamide

  • MDL number MFCD00684398
  • Cas Number : 55816-01-8
  • Molecular Weight : 295.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8807
Availability details for
Quantity
  • Availability
  • Add to Cart