COMH93D5CDB9 Combi-Blocks, Inc.

N-(4-Cyanophenyl)-4-methylbenzenesulfonamide

  • MDL number MFCD01006537
  • Cas Number : 56768-53-7
  • Molecular Weight : 272.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8927
Availability details for
Quantity
  • Availability
  • Add to Cart