COMH93D5CDD5 Combi-Blocks, Inc.

2-Bromo-4-chloro-1-methyl-1h-imidazo[4,5-c]pyridine

  • MDL number MFCD08062997
  • Cas Number : 496955-66-9
  • Molecular Weight : 246.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8970

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8970
Availability details for
Quantity
  • Availability
  • Add to Cart