COMH93D5CE18 Combi-Blocks, Inc.

1-(2,5-Dichlorophenyl)-2-nitroethene

  • MDL number MFCD03410754
  • Cas Number : 1056473-63-2
  • Molecular Weight : 218.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9074
Availability details for
Quantity
  • Availability
  • Add to Cart