COMH93D5CE1C Combi-Blocks, Inc.

1-(3-Chlorobenzyl)-3-methyl-1h-pyrazol-5-amine

  • MDL number MFCD08457372
  • Cas Number : 1006682-91-2
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9085
Availability details for
Quantity
  • Availability
  • Add to Cart