COMH93D5CE5B Combi-Blocks, Inc.

6-Carboxytetramethylrhodamine n-hydroxysuccinimide ester

  • MDL number MFCD00467520
  • Cas Number : 150810-69-8
  • Molecular Weight : 527.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9188
Availability details for
Quantity
  • Availability
  • Add to Cart