COMH93D5CE75 Combi-Blocks, Inc.

5-Methyl-l-uridine

  • MDL number MFCD02683607
  • Cas Number : 642082-80-2
  • Molecular Weight : 258.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9227
Availability details for
Quantity
  • Availability
  • Add to Cart