COMH93D5CE77 Combi-Blocks, Inc.

4-Bromo-2,6-dimethylaniline hydrobromide

  • MDL number MFCD08276249
  • Cas Number : 101252-37-3
  • Molecular Weight : 280.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9229
Availability details for
Quantity
  • Availability
  • Add to Cart