COMH93D5CE87 Combi-Blocks, Inc.

1-(3-Methoxyphenyl)-n-(2-thienylmethyl)methanamine

  • MDL number MFCD07364893
  • Cas Number : 880814-25-5
  • Molecular Weight : 233.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9257
Availability details for
Quantity
  • Availability
  • Add to Cart