COMH93D5CEE7 Combi-Blocks, Inc.

Schisandrol b

  • MDL number MFCD01941645
  • Cas Number : 58546-54-6
  • Molecular Weight : 416.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9407
Availability details for
Quantity
  • Availability
  • Add to Cart