COMH93D5CEF7 Combi-Blocks, Inc.

1-(3-Fluoro-4-methoxyphenyl)-2-nitropropene

  • MDL number MFCD01317634
  • Cas Number : 1023717-11-4
  • Molecular Weight : 211.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9426
Availability details for
Quantity
  • Availability
  • Add to Cart