COMH93D5CF20 Combi-Blocks, Inc.

Benzyl 11-bromoundecanoate

  • MDL number MFCD00017890
  • Cas Number : 78277-30-2
  • Molecular Weight : 355.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9494
Availability details for
Quantity
  • Availability
  • Add to Cart