COMH93D5CF2F Combi-Blocks, Inc.

2-(Trifluoromethyl)indolizine-1-carboxylic acid ethyl ester

  • MDL number MFCD08447583
  • Cas Number : 1284226-15-8
  • Molecular Weight : 257.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9517
Availability details for
Quantity
  • Availability
  • Add to Cart