COMH93D5CF43 Combi-Blocks, Inc.

4-(Chloromethyl)-2-(3-chlorophenyl)thiazole hydrochloride

  • MDL number MFCD05664422
  • Cas Number : 690632-83-8
  • Molecular Weight : 280.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9548
Availability details for
Quantity
  • Availability
  • Add to Cart