COMH93D5CF50 Combi-Blocks, Inc.

2-[4-(Thiophen-2-yl)phenyl]ethan-1-amine

  • MDL number MFCD08449280
  • Cas Number : 910400-56-5
  • Molecular Weight : 203.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9566
Availability details for
Quantity
  • Availability
  • Add to Cart