COMH93D5CF5C Combi-Blocks, Inc.

4'-(2-Hydroxyethoxy)-3'-nitroacetanilide

  • MDL number MFCD07783661
  • Cas Number : 59820-33-6
  • Molecular Weight : 240.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9582
Availability details for
Quantity
  • Availability
  • Add to Cart