COMH93D5D0BC Combi-Blocks, Inc.

5-Bromo-3-ethyl-1-benzothiophene

  • MDL number MFCD09038050
  • Cas Number : 900508-78-3
  • Molecular Weight : 241.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0100
Availability details for
Quantity
  • Availability
  • Add to Cart