COMH93D5D0FA Combi-Blocks, Inc.

4-(Acetylamino)-2-fluorobenzenesulfonyl chloride

  • MDL number MFCD09043386
  • Cas Number : 344-70-7
  • Molecular Weight : 251.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0170
Availability details for
Quantity
  • Availability
  • Add to Cart