COMH93D5D147 Combi-Blocks, Inc.

3-Phenyl-1h-indazol-5-amine

  • MDL number MFCD09261133
  • Cas Number : 395099-05-5
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0253
Availability details for
Quantity
  • Availability
  • Add to Cart