COMH93D5D170 Combi-Blocks, Inc.

Wst-5

  • MDL number MFCD09264686
  • Cas Number : 178925-55-8
  • Molecular Weight : 1331.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0302
Availability details for
Quantity
  • Availability
  • Add to Cart